missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylmorpholine N-oxide, 50 wt.% solution in water
CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol
$348.96 - $348.96
Specifications
Chemical Name or Material | 4-Methylmorpholine N-oxide |
---|---|
Name Note | 50 wt.% solution in water |
CAS | 7529-22-8,7732-18-5 |
CAS Min % | 50.0,48.0 |
CAS Max % | 52.0,50.0 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC348270010
|
Thermo Scientific Chemicals
348270010 |
1 kg | Glass bottle |
Each for $348.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
4-Methylmorpholine N-oxide | |
7529-22-8,7732-18-5 | |
52.0,50.0 | |
Colorless to Yellow | |
118.0°C to 119.0°C | |
Glass bottle | |
MFCD00005947 | |
15, 6171 | |
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f | |
LFTLOKWAGJYHHR-UHFFFAOYSA-N | |
4-methyl-4-oxidomorpholin-4-ium | |
82029 | |
117.15 |
50 wt.% solution in water | |
50.0,48.0 | |
-20.0°C | |
1.1300g/mL | |
>100°C | |
C5H11NO2 | |
07,244 | |
1.13 | |
Solubility in water: miscible | |
C[N+]1([O-])CCOCC1 | |
117.15 | |
CHEBI:52093 | |
Liquid |
RUO – Research Use Only