missing translation for 'onlineSavingsMsg'
Learn More

4-Methylaminophenol sulfate, 99%, extra pure

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning | CAS: 55-55-0 | C14H18N2O2·H2SO4 | 344.38 g/mol

Supplier:  Thermo Scientific Chemicals 156805000

 View more versions of this product

Catalog No. AC156805000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure if swallowed. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health: 2
Flammability: 1
Instability: 1

GHS Signal Word: Warning

EINECSNumber : 200-237-1

RTECSNumber : SL8650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only