Learn More
4-Methyl-2-pentanone, 99.5%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 255660250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Recyclable
Specifications
4-Methyl-2-pentanone | |
0.0004 meq/g max. | |
100.0 | |
117.4°C | |
Authentic | |
Metal drum | |
1.3950 to 1.3970 | |
MFCD00008938 | |
01,691 | |
0.8 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
100.16 | |
Analysis |
108-10-1 | |
99.4 | |
0.8000g/mL | |
14°C | |
99.4% min. (GC) | |
C6H12O | |
(CH3)2CHCH2COCH3 | |
25L | |
0.005% max. | |
15,5253 | |
Solubility in water: 17g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride and acetone | |
CC(C)CC(=O)C | |
100.16 | |
CHEBI:82344 | |
99.5% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-550-1
RUO â Research Use Only