Learn More
4'-Methoxyacetophenone, 98%
CAS: 100-06-1 | C9H10O2 | 150.18 g/mol
Supplier: Thermo Scientific Chemicals 152360010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4'-Methoxyacetophenone | |
100-06-1 | |
100.0 | |
White | |
138°C | |
97.5% min. (GC) | |
C9H10O2 | |
MFCD00008745 | |
08, 87 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
COC1=CC=C(C=C1)C(C)=O | |
150.18 | |
CHEBI:86567 | |
98% |
98% | |
97.5 | |
36.0°C to 39.0°C | |
260.0°C | |
Authentic | |
Glass bottle | |
CH3OC6H4COCH3 | |
1 kg | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)ethan-1-one | |
7476 | |
150.18 | |
Crystalline Powder or Crystals |
Chemical Identifiers
100-06-1 | |
150.18 | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
7476 | |
COC1=CC=C(C=C1)C(C)=O |
C9H10O2 | |
MFCD00008745 | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenylShow More | |
CHEBI:86567 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 202-815-9
RTECSNumber : AM9240000
TSCA : TSCA
RUO – Research Use Only