Learn More
4-Methoxy-3-buten-2-one, 90%, tech.

CAS: 4652-27-1 | C5H8O2 | 100.12 g/mol
$125.58 - $125.58
Chemical Identifiers
CAS | 4652-27-1 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00008778,MFCD00008778,MFCD00008778 |
InChI Key | VLLHEPHWWIDUSS-ARJAWSKDSA-N |
Synonym | z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one |
PubChem CID | 643837 |
IUPAC Name | (Z)-4-methoxybut-3-en-2-one |
SMILES | CO\C=C/C(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC153390100
|
Thermo Scientific Chemicals
153390100 |
10 mL | Glass bottle |
Each for $125.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4652-27-1 | |
100.12 | |
VLLHEPHWWIDUSS-ARJAWSKDSA-N | |
643837 | |
CO\C=C/C(C)=O |
C5H8O2 | |
MFCD00008778,MFCD00008778,MFCD00008778 | |
z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one | |
(Z)-4-methoxybut-3-en-2-one |
Specifications
4652-27-1 | |
100.0 | |
0.9800g/mL | |
63°C | |
88% min. (GC) | |
C5H8O2 | |
CH3OCH=CHCOCH3 | |
10 mL | |
10,258 | |
z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one | |
VLLHEPHWWIDUSS-ARJAWSKDSA-N | |
(Z)-4-methoxybut-3-en-2-one | |
643837 | |
90% | |
Liquid |
88.0 | |
Yellow | |
200°C | |
Authentic | |
Glass bottle | |
1.4650 to 1.469 | |
MFCD00008778,MFCD00008778,MFCD00008778 | |
01, IV, 4080 | |
0.98 | |
Solubility in water: insoluble | |
CO\C=C/C(C)=O | |
100.12 | |
100.12 | |
Technical | |
4-Methoxy-3-buten-2-one, tech., 90% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 225-087-4
RUO – Research Use Only