missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI Americaâ„¢
$205.98 - $710.56
Chemical Identifiers
CAS | 15051-46-4 |
---|---|
Molecular Formula | C13H22NO3 |
Molecular Weight (g/mol) | 240.32 |
MDL Number | MFCD17677357 |
InChI Key | BTWSPOZXDCFMLX-UHFFFAOYSA-N |
Synonym | 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer |
PubChem CID | 9859588 |
IUPAC Name | {2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl |
SMILES | CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1 |
Chemical Identifiers
15051-46-4 | |
240.32 | |
BTWSPOZXDCFMLX-UHFFFAOYSA-N | |
9859588 | |
CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1 |
C13H22NO3 | |
MFCD17677357 | |
4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer | |
{2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl |
Specifications
15051-46-4 | |
C13H22NO3 | |
1 g | |
BTWSPOZXDCFMLX-UHFFFAOYSA-N | |
{2,2,6,6-tetramethyl-4-[(2-methylprop-2-enoyl)oxy]piperidin-1-yl}oxidanyl | |
9859588 | |
≥98.0% (GC,T) | |
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
89°C | |
MFCD17677357 | |
4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer | |
CC(=C)C(=O)OC1CC(C)(C)N([O])C(C)(C)C1 | |
240.32 | |
240.32 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator