missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenoxymethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (1.0-2.0mmol/g), TCI Americaâ„¢

$257.24 - $257.24

Chemical Identifiers

MDL Number MFCD00131778
Synonym Wang Resin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09875G
View Documents
TCI America
H09875G
5 g
Each for $257.24
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00131778
Wang Resin

Specifications

MFCD00131778
Wang Resin
4-(Hydroxymethyl)phenoxymethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (1.0-2.0mmol
5 g
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'