Learn More
4-Hydroxy-3,6-dimethyl-2-pyrone, 97%
CAS: 5192-62-1 | C7H8O3 | 140.138 g/mol
$282.45 - $1048.59
Chemical Identifiers
| CAS | 5192-62-1 |
|---|---|
| Molecular Formula | C7H8O3 |
| Molecular Weight (g/mol) | 140.138 |
| MDL Number | MFCD00090424 |
| InChI Key | VVBIGJOVPZMWGU-UHFFFAOYSA-N |
| Synonym | 4-hydroxy-3,6-dimethyl-2h-pyran-2-one, 3,6-dimethyl-4-hydroxy-2-pyrone, chembl51918, 2h-pyran-2-one, 4-hydroxy-3,6-dimethyl, 2-hydroxy-3,6-dimethylpyran-4-one, hdmpo, 2-hydroxy-3,6-dimethyl-4h-pyran-4-one, acmc-20amh7, methyl triacetic lactone, 4-hydroxy-3,6-dimethyl-2-pyrone |
| PubChem CID | 54690337 |
| IUPAC Name | 4-hydroxy-3,6-dimethylpyran-2-one |
| SMILES | CC1=CC(=C(C(=O)O1)C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2607403
|
Thermo Scientific Chemicals
H2607403 |
1 g |
Each for $282.45
|
|
|||||
|
AAH2607406
|
Thermo Scientific Chemicals
H2607406 |
5 g |
Each for $1,048.59
|
|
|||||
Description
4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxy-3,6-dimethyl-2-pyrone is the reactant in the formation of unnatural novel polyketides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 5192-62-1 | |
| 140.138 | |
| VVBIGJOVPZMWGU-UHFFFAOYSA-N | |
| 54690337 | |
| CC1=CC(=C(C(=O)O1)C)O |
| C7H8O3 | |
| MFCD00090424 | |
| 4-hydroxy-3,6-dimethyl-2h-pyran-2-one, 3,6-dimethyl-4-hydroxy-2-pyrone, chembl51918, 2h-pyran-2-one, 4-hydroxy-3,6-dimethyl, 2-hydroxy-3,6-dimethylpyran-4-one, hdmpo, 2-hydroxy-3,6-dimethyl-4h-pyran-4-one, acmc-20amh7, methyl triacetic lactone, 4-hydroxy-3,6-dimethyl-2-pyrone | |
| 4-hydroxy-3,6-dimethylpyran-2-one |
Specifications
| 5192-62-1 | |
| C7H8O3 | |
| 1 g | |
| VVBIGJOVPZMWGU-UHFFFAOYSA-N | |
| 4-hydroxy-3,6-dimethylpyran-2-one | |
| 54690337 | |
| 97% |
| 209°C to 211°C | |
| MFCD00090424 | |
| 4-hydroxy-3,6-dimethyl-2h-pyran-2-one, 3,6-dimethyl-4-hydroxy-2-pyrone, chembl51918, 2h-pyran-2-one, 4-hydroxy-3,6-dimethyl, 2-hydroxy-3,6-dimethylpyran-4-one, hdmpo, 2-hydroxy-3,6-dimethyl-4h-pyran-4-one, acmc-20amh7, methyl triacetic lactone, 4-hydroxy-3,6-dimethyl-2-pyrone | |
| CC1=CC(=C(C(=O)O1)C)O | |
| 140.138 | |
| 140.14 | |
| 4-Hydroxy-3,6-dimethyl-2-pyrone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only