missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 189681-04-7 |
---|---|
Molecular Formula | C10H5ClN2S2 |
Molecular Weight (g/mol) | 252.73 |
MDL Number | MFCD00174019 |
InChI Key | VNWBUFWVNCUUKY-UHFFFAOYSA-N |
Synonym | 4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine |
PubChem CID | 735732 |
IUPAC Name | 4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine |
SMILES | ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AW00007DA
|
Thermo Scientific Chemicals
AW00007DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AW00007EA
|
Thermo Scientific Chemicals
AW00007EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
189681-04-7 | |
252.73 | |
VNWBUFWVNCUUKY-UHFFFAOYSA-N | |
735732 | |
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1 |
C10H5ClN2S2 | |
MFCD00174019 | |
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine | |
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine |
Specifications
189681-04-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD00174019 | |
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine | |
ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1 | |
252.73 | |
252.75 | |
4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine |
94.5 | |
95% | |
C10H5ClN2S2 | |
1g | |
VNWBUFWVNCUUKY-UHFFFAOYSA-N | |
4-chloro-5-(thiophen-2-yl)thieno[2,3-d]pyrimidine | |
735732 | |
95% |