missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-2-thienyl)methanol, 97%, Thermo Scientific™
GREENER_CHOICE

$63.51 - $63.51

Chemical Identifiers

CAS 79757-77-0
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.058
MDL Number MFCD04115392
InChI Key PXZNJHHUYJRFPZ-UHFFFAOYSA-N
PubChem CID 2795484
IUPAC Name (4-bromothiophen-2-yl)methanol
SMILES C1=C(SC=C1Br)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC28509DA
SDS View Documents
Thermo Scientific Chemicals
CC28509DA
1 g Amber Glass Bottle
Each for $63.51
Only null left
 
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'