missing translation for 'onlineSavingsMsg'
Learn More

4-(4-chlorophenyl)piperidin-4-ol, 97%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 39512-49-7
Molecular Formula C11H15Cl2NO
Molecular Weight (g/mol) 248.15
MDL Number MFCD00006001
InChI Key PXAJMBHRLGKPQV-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine
PubChem CID 38282
IUPAC Name 4-(4-chlorophenyl)piperidin-4-ol
SMILES [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09284EA
SDS View Documents
Thermo Scientific Chemicals
BTB09284EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09284EB
SDS View Documents
Thermo Scientific Chemicals
BTB09284EB
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09284FA
SDS View Documents
Thermo Scientific Chemicals
BTB09284FA
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'