missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC366380050

 View more versions of this product

Catalog No. AC366380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Glass bottle
165°C to 170°C
97%
96.0
96% min. (GC)
MFCD02093721
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N
219.09
219.09
97%
Authentic
Beige to White
5g
214360-73-3
100.0
C12H18BNO2
2-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 4-aminobenzeneboronic acid pinacol ester, 4-aminophenylboronic acid pinacol ester, 4-aminophenylboronic acid pinacolate, 4-aminophenylboronic acid, pinacol ester, 4-aminophenylboronicacidpinacolester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, pinacol cyclic ester
ZANPJXNYBVVNSD-UHFFFAOYSA-N
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
2734620
Crystalline Powder

Chemical Identifiers

214360-73-3
219.09
ZANPJXNYBVVNSD-UHFFFAOYSA-N
2734620
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N
C12H18BNO2
MFCD02093721
2-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 4-aminobenzeneboronic acid pinacol ester, 4-aminophenylboronic acid pinacol ester, 4-aminophenylboronic acid pinacolate, 4-aminophenylboronic acid, pinacol ester, 4-aminophenylboronicacidpinacolester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, pinacol cyclic ester
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : product may turn dark on exposure to air

SDS Documents