missing translation for 'onlineSavingsMsg'
Learn More

4,4,4,4',4',4'-Hexafluoro-DL-valine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 446540050

 View more versions of this product

Catalog No. AC446540050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

4,4,4,4',4',4'-Hexafluoro-DL-valine
96 to 104%
MFCD00014349
Solubility in water: soluble. Other solubilities: soluble in mineral acids
NC(C(C(F)(F)F)C(F)(F)F)C(O)=O
225.09
97%
16063-80-2
C5H5F6NO2
5g
KRNSHCKTGFAMPQ-UHFFFAOYNA-N
2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
225.09

Chemical Identifiers

16063-80-2
225.09
KRNSHCKTGFAMPQ-UHFFFAOYNA-N
NC(C(C(F)(F)F)C(F)(F)F)C(O)=O
C5H5F6NO2
MFCD00014349
2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 240-207-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only