missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ N95 and P95 Particulate Filters (5000/6000 Series)
NIOSH approved for environments containing certain non-oil based particles. Use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501.
$30.66 - $41.19
Specifications
Cartridge Type | Filter Cartridge |
---|---|
Certifications/Compliance | NIOSH |
For Use With (Equipment) | 3M™ respirator 5000 series or 3M™ cartridge 6000 series or 3M™ filter adapter 603 and 3M™ filter retainer 501 |
Length (English) | 4.2 in. |
Length (Metric) | 10.6 cm |
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Price | Quantity | |||||
18-999-4547
|
3M™
7000002008 |
N95 Particulate Filter | Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding |
Pack of 10 Each for $30.66
Case of 10 Pack for $287.40
|
|
|||||
19-002717
|
3M™
7000002054 |
P95 Particulate Filter | Cleaning, demolition, facility maintenance, grinding, painting, sanding, welding |
Pack of 10 Each for $41.19
Case of 10 Pack for $67.50
|
|
|||||
Description
The 3M™ Particulate Filter 5N11, N95 helps provide respiratory protection in environments containing certain non-oil based particles. When properly fitted, use with appropriate cartridges for various applications including spray painting, petrochemical and chemical manufacturing, and pesticide spraying for concentrations up to 10 times the Permissible Exposure Limit (PEL) with half facepieces, or 50 times PEL with quantitatively fit tested full facepieces. Filter protects against: 1) solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances; 2) liquids or oil based particles from sprays that do not emit harmful vapors; and 3) metal fumes produced from welding, brazing, cutting, and other operations involving heating of metals.NIOSH (National Institute for Occupational Safety and Health) approved for environments containing certain non-oil based particles. This filter has been rated N95, which means it filters at least 95% of airborne particles, but is not resistant to oil.Breathing hazardous particles can pose a risk to your health. NIOSH, a Federal government regulatory agency, has tested and approved the 3M™ Particulate Filter 5N11 to help reduce exposure to certain particles.- NIOSH approved for environments containing certain non-oil-based particles
- Replaceable particle filter for cost savings
- Wide range of applications reduces inventory needs and training requirements
- Designed for use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501
- 3M Catalog# 5N11 , 3M Catalog# 5P71
Specifications
Filter Cartridge | |
3M™ respirator 5000 series or 3M™ cartridge 6000 series or 3M™ filter adapter 603 and 3M™ filter retainer 501 | |
10.6 cm | |
8.6 cm |
NIOSH | |
4.2 in. | |
3.4 in. |