Learn More
3M™ Disposable N95 Filtering Welding and Metal Pouring Respirator
For use when welding, torch cutting, soldering, brazing and working with molten metals
Supplier:  3M™ 70070799534
Catalog No. 29-612-332
Only null left
Explore available promotions
Alert:
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Description
Description
3M Filtering Facepiece Particulate Respirators (FFRs) meet NIOSH 42CFR84 standards. Feature Advanced Electret Media designed for ease of breathing. Recommended for use up to 10X the OSHA PEL or applicable occupational exposure limits, whichever is lower.
Application
  • For use when welding, torch cutting, soldering, brazing, and working with molten metals
  • Design
    • All styles feature Cool Flow™ exhalation valve
    • All except 19-038-989 additionally feature adjustable buckle straps and cake-resistant filter media for comfort and durability
    • Available with or without a chemical absorbent for relief from nuisance level welding odors
    • Unique filter media incorporating protective layers designed to be flame resistant as demonstrated per modified ASTM™ D2859-06 (Reapproved 2011)
    • 3M Catalog# 8212, 8214, 8512, 8514, 8515, 8516
    Compliance
    Meets NIOSH 42 CFR 84 N95 requirements. NIOSH Approval No.: TC-84A-0454.
Product Suggestions
Product Suggestions