Learn More
3-n-Hexylthiophene, 98%
CAS: 1693-86-3 | C10H16S | 168.298 g/mol
Supplier: Thermo Scientific Chemicals H5589103
Description
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
Solubility
Partly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
- Environmental benefits include:
- Renewable Energy
Specifications
3-n-Hexylthiophene | |
0.936 | |
37°C (99°F) | |
C10H16S | |
MFCD00143181 | |
UN1993 | |
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+% | |
JEDHEMYZURJGRQ-UHFFFAOYSA-N | |
3-hexylthiophene | |
566849 | |
≥99% |
1693-86-3 | |
65°C (0.45 mmHg) | |
Stench | |
1.496 | |
1 g | |
1617129 | |
Partly soluble in water. | |
CCCCCCC1=CSC=C1 | |
168.298 | |
168.3 |
Chemical Identifiers
1693-86-3 | |
168.298 | |
JEDHEMYZURJGRQ-UHFFFAOYSA-N | |
566849 | |
CCCCCCC1=CSC=C1 |
C10H16S | |
MFCD00143181 | |
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%Show More | |
3-hexylthiophene |
Safety and Handling
GHS H Statement
H301-H311-H226-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only