Learn More
3-Mercapto-1,2-propanediol, 90 wt% aqueous solution
CAS: 96-27-5 | C3H8O2S | 108.16 g/mol
$111.90 - $111.90
Chemical Identifiers
CAS | 96-27-5 |
---|---|
Molecular Formula | C3H8O2S |
Molecular Weight (g/mol) | 108.16 |
MDL Number | MFCD00004879 |
InChI Key | PJUIMOJAAPLTRJ-UHFFFAOYNA-N |
Synonym | 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiolShow More |
PubChem CID | 7291 |
ChEBI | CHEBI:74537 |
IUPAC Name | 3-sulfanylpropane-1,2-diol |
SMILES | OCC(O)CS |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC125520250
|
Thermo Scientific Chemicals
125520250 |
25 mL | Glass bottle |
Each for $111.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-27-5 | |
108.16 | |
PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
7291 | |
3-sulfanylpropane-1,2-diol |
C3H8O2S | |
MFCD00004879 | |
3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiolShow More | |
CHEBI:74537 | |
OCC(O)CS |
Specifications
96-27-5,7732-18-5 | |
92.0,12.0 | |
1.2300g/mL | |
>112°C | |
Glass bottle | |
1.5073 | |
MFCD00004879 | |
01,519 | |
15,9488 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,insoluble in ether | |
OCC(O)CS | |
108.16 | |
CHEBI:74537 | |
≥88% | |
3-Mercapto-1, 2-propanediol |
88.0,8.0 | |
Colorless to Yellow | |
118.0°C (5.0 mmHg) | |
Authentic | |
C3H8O2S | |
HSCH2CH(OH)CH2OH | |
25 mL | |
1.23 | |
3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
3-sulfanylpropane-1,2-diol | |
7291 | |
108.16 | |
Viscous Solution |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get
GHS Signal Word: Danger
EINECSNumber : 202-495-0
RTECSNumber : TY8140000
TSCA : TSCA
RUO – Research Use Only