missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Heptanone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 120380025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
3-Heptanone | |
0.8100g/mL | |
41°C | |
98% | |
C7H14O | |
CH3(CH2)3COC2H5 | |
2.5L | |
0.81 | |
Solubility in water: 3.3g/L water (20° c). Other solubilities: soluble in alcohol and ether | |
CCCCC(=O)CC | |
114.19 | |
CHEBI:50139 | |
98% |
106-35-4 | |
146.0°C to 149.0°C | |
Authentic | |
Glass bottle | |
1.4075 to 1.4095 | |
MFCD00009483 | |
01,699 | |
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone | |
NGAZZOYFWWSOGK-UHFFFAOYSA-N | |
heptan-3-one | |
7802 | |
114.19 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 203-388-1
RUO â Research Use Only