Learn More
3,5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 175135-06-5 |
---|---|
Molecular Formula | C12H5Cl4NO5S |
Molecular Weight (g/mol) | 417.034 |
MDL Number | MFCD00052029 |
InChI Key | YNPAAILIMIGQIH-UHFFFAOYSA-N |
Synonym | 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy |
PubChem CID | 2774361 |
IUPAC Name | 3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BTB03983DA
|
Thermo Scientific Chemicals
BTB03983DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB03983EA
|
Thermo Scientific Chemicals
BTB03983EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
175135-06-5 | |
417.034 | |
YNPAAILIMIGQIH-UHFFFAOYSA-N | |
2774361 | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl |
C12H5Cl4NO5S | |
MFCD00052029 | |
3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy | |
3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride |
Specifications
175135-06-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD00052029 | |
3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl | |
417.034 | |
417.05 | |
3, 5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride |
96.5 | |
97% | |
C12H5Cl4NO5S | |
1g | |
YNPAAILIMIGQIH-UHFFFAOYSA-N | |
3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride | |
2774361 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger