missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',5,7-Trihydroxy-4'-methoxyflavanone, 97%

Manufacturer:  Alfa Aesar™ B2052814

 View more versions of this product

Catalog No. AAB2052814



Description

Description

Applications
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

3',5,7-Trihydroxy-4'-methoxyflavanone
25g
C16H14O6
309850
--hesperetin, 3',5,7-trihydroxy-4'-methoxyflavanone, 5,7,3'-trihydroxy-4'-methoxyflavanone, cyanidanon 4'-methyl ether 1626, hesperetin, hesperin, hesperitin, prestwick_908, unii-q9q3d557f1, yso2
AIONOLUJZLIMTK-AWEZNQCLSA-N
(2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
72281
302.3
227°C to 232°C
520-33-2
MFCD00075646
14,4670
Soluble in water (partly),dilute alkalis,and ethanol (50mg/ml).
COC1=C(C=C(C=C1)C2CC(=O)C3=C(C=C(C=C3O2)O)O)O
302.282
CHEBI:28230
97%

Chemical Identifiers

520-33-2
302.282
AIONOLUJZLIMTK-AWEZNQCLSA-N
72281
(2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
C16H14O6
MFCD00075646
--hesperetin, 3',5,7-trihydroxy-4'-methoxyflavanone, 5,7,3'-trihydroxy-4'-methoxyflavanone, cyanidanon 4'-methyl ether 1626, hesperetin, hesperin, hesperitin, prestwick_908, unii-q9q3d557f1, yso2
CHEBI:28230
COC1=C(C=C(C=C1)C2CC(=O)C3=C(C=C(C=C3O2)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 208-290-2

TSCA : No

Recommended Storage : Ambient temperatures

Documents