missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 20825-89-2 |
---|---|
Molecular Formula | C10H10O4 |
Molecular Weight (g/mol) | 194.186 |
MDL Number | MFCD00276310 |
InChI Key | MQSSBVLREFSMDP-UHFFFAOYSA-N |
Synonym | 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro |
PubChem CID | 2776383 |
IUPAC Name | 3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid |
SMILES | C1COC2=C(C=C(C=C2)C(=O)O)OC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC13201CB
|
Thermo Scientific Chemicals
CC13201CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC13201DA
|
Thermo Scientific Chemicals
CC13201DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
20825-89-2 | |
194.186 | |
MQSSBVLREFSMDP-UHFFFAOYSA-N | |
2776383 | |
C1COC2=C(C=C(C=C2)C(=O)O)OC1 |
C10H10O4 | |
MFCD00276310 | |
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid |
Specifications
20825-89-2 | |
100.0 | |
Amber Glass Bottle | |
MFCD00276310 | |
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro | |
C1COC2=C(C=C(C=C2)C(=O)O)OC1 | |
194.186 | |
194.19 | |
3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carboxylic acid |
96.5 | |
97% | |
C10H10O4 | |
250mg | |
MQSSBVLREFSMDP-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid | |
2776383 | |
97% |