missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethylglutaric acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116241000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
3, 3-Dimethylglutaric acid | |
97.5 | |
290.0°C | |
98% | |
C7H12O4 | |
MFCD00002716 | |
02, 684 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions | |
CC(C)(CC(O)=O)CC(O)=O | |
160.17 | |
CHEBI:68502 | |
98% |
4839-46-7 | |
100.0 | |
Authentic | |
Glass bottle | |
HO2CCH2C(CH3)2CH2CO2H | |
100g | |
3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
3,3-dimethylpentanedioic acid | |
20984 | |
160.17 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 225-425-
RUO â Research Use Only