missing translation for 'onlineSavingsMsg'
Learn More

2-(Propylthio)pyridine-3-carbonyl chloride, 97%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 175135-24-7
Molecular Formula C9H10ClNOS
Molecular Weight (g/mol) 215.70
MDL Number MFCD00052656
InChI Key FTEUTJYYROCUBI-UHFFFAOYSA-N
Synonym 2-propylthio pyridine-3-carbonyl chloride, 2-propylsulfanyl pyridine-3-carbonyl chloride, 2-propylthio nicotinoyl chloride, 2-propylthiopyridine-3-carbonyl chloride, 3-pyridinecarbonylchloride, 2-propylthio, 2-propylthio pyridine-3-carbonylchlorideShow MoreShow Less
PubChem CID 2774530
IUPAC Name 2-propylsulfanylpyridine-3-carbonyl chloride
SMILES CCCSC1=C(C=CC=N1)C(Cl)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB06371DA
SDS View Documents
Thermo Scientific Chemicals
BTB06371DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB06371EA
SDS View Documents
Thermo Scientific Chemicals
BTB06371EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'