Learn More
2-Nitrophenyl isocyanate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 296690250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Recyclable
Specifications
2-Nitrophenyl isocyanate | |
98.5 | |
135.0°C to 137.0°C (17.0mmHg) | |
Authentic | |
Glass bottle | |
O2NC6H4NCO | |
2-nitrophenyl isocyanate, o-nitrophenyl isocyanate, benzene, 1-isocyanato-2-nitro, isocyanic acid, o-nitrophenyl ester, 2-nitrophenylisocyanate, 2-nitrobenzenisocyanate, nitrophenyl isocyanate, 2-nitrophenylisocynate, 2-nitro phenyl isocyanate, acmc-1cp5k | |
JRVZITODZAQRQM-UHFFFAOYSA-N | |
1-isocyanato-2-nitrobenzene | |
76834 | |
99% |
3320-86-3 | |
100.0 | |
>110°C | |
98.5% min. (GC) | |
C7H4N2O3 | |
25g | |
Solubility in water: decomposes. Other solubilities: soluble in toluene | |
C1=CC=C(C(=C1)N=C=O)[N+](=O)[O-] | |
164.12 | |
164.12 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 222-024-2
RUO â Research Use Only