Learn More
2-Methyl-5-phenyl-3-furoic acid, 97%, Thermo Scientific™

$91.13 - $91.13
Chemical Identifiers
CAS | 108124-17-0 |
---|---|
Molecular Formula | C12H9O3 |
Molecular Weight (g/mol) | 201.20 |
MDL Number | MFCD00221070 |
InChI Key | VLMNACSEESRUAK-UHFFFAOYSA-M |
Synonym | 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acidShow More |
PubChem CID | 736516 |
IUPAC Name | 2-methyl-5-phenylfuran-3-carboxylic acid |
SMILES | CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O |
Chemical Identifiers
108124-17-0 | |
201.20 | |
VLMNACSEESRUAK-UHFFFAOYSA-M | |
736516 | |
CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O |
C12H9O3 | |
MFCD00221070 | |
2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acidShow More | |
2-methyl-5-phenylfuran-3-carboxylic acid |
Specifications
108124-17-0 | |
100.0 | |
Amber Glass Bottle | |
MFCD00221070 | |
2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid | |
CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O | |
201.20 | |
202.21 | |
2-Methyl-5-phenyl-3-furoic acid |
96.5 | |
97% | |
C12H9O3 | |
1 g | |
VLMNACSEESRUAK-UHFFFAOYSA-M | |
2-methyl-5-phenylfuran-3-carboxylic acid | |
736516 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.