missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 1034566-13-6
Molecular Formula C6H4F3NOS
Molecular Weight (g/mol) 195.159
MDL Number MFCD12198128
InChI Key FBQFBBGXQZDOQV-UHFFFAOYSA-N
Synonym 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carbaldehyde, 5-formyl-2-methyl-4-trifluoromethyl-1,3-thiazole
PubChem CID 43811060
IUPAC Name 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
SMILES CC1=NC(=C(S1)C=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC76304CB
SDS View Documents
Thermo Scientific Chemicals
CC76304CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC76304DA
SDS View Documents
Thermo Scientific Chemicals
CC76304DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'