missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-2-methyloxirane, 97%, Thermo Scientificâ„¢

$566.53 - $566.53

Chemical Identifiers

CAS 598-09-4
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00052487
InChI Key VVHFXJOCUKBZFS-UHFFFAOYSA-N
Synonym 2-chloromethyl-2-methyloxirane, oxirane, 2-chloromethyl-2-methyl, 2-chloromethyl-2-methyl-oxirane, 2-chloromethyl-1,2-epoxypropane, methyl epichlorohydrin, 2-methylepichlorohydrin, acmc-209mfx, beta-methylepichlorohydrin, ksc493o4b
PubChem CID 95220
IUPAC Name 2-(chloromethyl)-2-methyloxirane
SMILES CC1(CO1)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CD08593EA
SDS View Documents
Thermo Scientific Chemicals
CD08593EA
10 g Amber Glass Bottle
Each for $566.53
Only null left
 
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'