Learn More
2-Bromo-3'-chloropropiophenone, 98%
CAS: 34911-51-8 | C9H8BrClO | 247.52 g/mol
$67.11 - $238.03
Chemical Identifiers
CAS | 34911-51-8 |
---|---|
Molecular Formula | C9H8BrClO |
Molecular Weight (g/mol) | 247.52 |
MDL Number | MFCD03701150 |
InChI Key | OFNMQTRHMBQQEA-UHFFFAOYNA-N |
PubChem CID | 4992116 |
IUPAC Name | 2-bromo-1-(3-chlorophenyl)propan-1-one |
SMILES | CC(Br)C(=O)C1=CC(Cl)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6499903
|
Thermo Scientific Chemicals
H6499903 |
1 g |
Each for $67.11
|
|
||||||
AAH6499906
|
Thermo Scientific Chemicals
H6499906 |
5 g |
Each for $238.03
|
|
||||||
Description
It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in acetonitrile, chloroform, dichloromethane and ethyl acetate.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
34911-51-8 | |
247.52 | |
OFNMQTRHMBQQEA-UHFFFAOYNA-N | |
2-bromo-1-(3-chlorophenyl)propan-1-one |
C9H8BrClO | |
MFCD03701150 | |
4992116 | |
CC(Br)C(=O)C1=CC(Cl)=CC=C1 |
Specifications
34911-51-8 | |
1.532 g/mL | |
1.577 | |
1 g | |
2-bromo-3'-chloropropiophenone, 2-bromo-1-3-chlorophenyl propan-1-one, 2-bromo-1-3-chlorophenyl-1-propanone, 3'-chloro-2-bromopropiophenone, 1-propanone,2-bromo-1-3-chlorophenyl, 1-propanone, 2-bromo-1-3-chlorophenyl, 2-bromo-1-3-chloro-phenyl-propan-1-one, bromochlorophenylpropanone, bromo-3'-chloropropiophenone, ksc573e4l | |
OFNMQTRHMBQQEA-UHFFFAOYNA-N | |
2-bromo-1-(3-chlorophenyl)propan-1-one | |
4992116 | |
98% | |
2-Bromo-3'-chloropropiophenone |
Colorless to Yellow | |
C9H8BrClO | |
MFCD03701150 | |
2360404 | |
Soluble in acetonitrile,chloroform,dichloromethane and ethyl acetate. | |
CC(Br)C(=O)C1=CC(Cl)=CC=C1 | |
247.52 | |
247.52 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 252-282-1
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only