Learn More
(±)-2-Amino-1-phenylethanol, 95%
CAS: 7568-93-6 | C8H11NO | 137.18 g/mol
$200.26 - $200.26
Chemical Identifiers
CAS | 7568-93-6 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.18 |
MDL Number | MFCD00008137 |
InChI Key | ULSIYEODSMZIPX-UHFFFAOYNA-N |
Synonym | phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanolShow More |
PubChem CID | 1000 |
ChEBI | CHEBI:16343 |
IUPAC Name | 2-amino-1-phenylethanol |
SMILES | NCC(O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC104320100
|
Thermo Scientific Chemicals
104320100 |
10 g | Glass bottle |
Each for $200.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7568-93-6 | |
137.18 | |
ULSIYEODSMZIPX-UHFFFAOYNA-N | |
1000 | |
2-amino-1-phenylethanol |
C8H11NO | |
MFCD00008137 | |
phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanolShow More | |
CHEBI:16343 | |
NCC(O)C1=CC=CC=C1 |
Specifications
7568-93-6 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
1.5580 to 1.5600 | |
MFCD00008137 | |
15, 7399 | |
ULSIYEODSMZIPX-UHFFFAOYNA-N | |
2-amino-1-phenylethanol | |
1000 | |
137.18 | |
Viscous Liquid |
97.5 | |
56.0°C to 60.0°C | |
160.0°C (17.0 mmHg) | |
97.5% min. (HPLC) | |
C8H11NO | |
H2NCH2CH(C6H5)OH | |
10 g | |
phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol | |
NCC(O)C1=CC=CC=C1 | |
137.18 | |
CHEBI:16343 | |
98% | |
(±)-2-Amino-1-phenylethanol, 98% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 231-469-1
RUO – Research Use Only