Learn More
2,7-Dibromonaphthalene, 99%
CAS: 58556-75-5 | C10H6Br2 | 285.97 g/mol
Supplier: Thermo Scientific Chemicals H5609706
Description
It is an intermediate in dyestuffs and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate in dyestuffs and pharmaceuticals.
Solubility
Soluble in acetonitrile.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
2,7-Dibromonaphthalene | |
139°C to 142°C | |
MFCD00238580 | |
UN3077 | |
naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc | |
ODJZWBLNJKNOJK-UHFFFAOYSA-N | |
2,7-dibromonaphthalene | |
625355 | |
99% |
58556-75-5 | |
C10H6Br2 | |
5 g | |
1862041 | |
Soluble in acetonitrile. | |
BrC1=CC2=CC(Br)=CC=C2C=C1 | |
285.97 | |
285.97 |
Chemical Identifiers
58556-75-5 | |
285.97 | |
ODJZWBLNJKNOJK-UHFFFAOYSA-N | |
625355 | |
BrC1=CC2=CC(Br)=CC=C2C=C1 |
C10H6Br2 | |
MFCD00238580 | |
naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc | |
2,7-dibromonaphthalene |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only