Learn More
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
CAS: 696-59-3 | C6H12O3 | 132.16 g/mol
Supplier: Thermo Scientific Chemicals 115671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 5-Dimethoxytetrahydrofuran | |
99% | |
Colorless to Yellow | |
100.0 | |
145°C to 148°C | |
Authentic | |
Liquid | |
99% | |
MFCD00005359 | |
2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
2,5-dimethoxyoxolane | |
79098 | |
99% |
-45°C | |
696-59-3 | |
98.5 | |
1.0210g/mL | |
100 g | |
Glass bottle | |
1.4160 to 1.419 | |
C6H12O3 | |
02,144 | |
Solubility in water: 350g/L (20°C). Other solubilities: miscible with alcohols and most common organic,solvents | |
COC1CCC(O1)OC | |
132.16 | |
132.16 |
Chemical Identifiers
696-59-3 | |
132.16 | |
GFISDBXSWQMOND-UHFFFAOYSA-N | |
79098 | |
COC1CCC(O1)OC |
C6H12O3 | |
MFCD00005359 | |
2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthfShow More | |
2,5-dimethoxyoxolane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INH
GHS Signal Word: Danger
EINECSNumber : 211-797-1
TSCA : TSCA
RUO – Research Use Only