Learn More
2,5-Dihydroxy-1,4-dithiane, 96%
CAS: 40018-26-6 | C4H8O2S2 | 152.226 g/mol
Supplier: Thermo Scientific Chemicals A146200B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,5-Dihydroxy-1,4-dithiane | |
∼130°C (decomposition) | |
MFCD00006659 | |
UN2811 | |
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme | |
C1C(SCC(S1)O)O | |
152.226 | |
152.24 |
40018-26-6 | |
C4H8O2S2 | |
1000 g | |
104402 | |
YUIOPHXTILULQC-UHFFFAOYSA-N | |
1,4-dithiane-2,5-diol | |
98330 | |
96% |
Chemical Identifiers
40018-26-6 | |
152.226 | |
YUIOPHXTILULQC-UHFFFAOYSA-N | |
98330 | |
C1C(SCC(S1)O)O |
C4H8O2S2 | |
MFCD00006659 | |
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anmeShow More | |
1,4-dithiane-2,5-diol |
Safety and Handling
GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
H301-H317-H318
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 254-751-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only