missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloro-5-pyrimidinecarboxaldehyde 98.0+%, TCI Americaâ„¢
$71.81 - $213.54
Chemical Identifiers
CAS | 50270-27-4 |
---|---|
Molecular Formula | C5HCl3N2O |
Molecular Weight (g/mol) | 211.426 |
MDL Number | MFCD02257700 |
InChI Key | KVJIRFGNHAAUNQ-UHFFFAOYSA-N |
Synonym | 2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehydeShow More |
PubChem CID | 10932746 |
IUPAC Name | 2,4,6-trichloropyrimidine-5-carbaldehyde |
SMILES | C(=O)C1=C(N=C(N=C1Cl)Cl)Cl |
Chemical Identifiers
50270-27-4 | |
211.426 | |
KVJIRFGNHAAUNQ-UHFFFAOYSA-N | |
10932746 | |
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl |
C5HCl3N2O | |
MFCD02257700 | |
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehydeShow More | |
2,4,6-trichloropyrimidine-5-carbaldehyde |
Specifications
50270-27-4 | |
White-Yellow | |
MFCD02257700 | |
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde | |
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl | |
211.426 | |
211.43 | |
Crystalline Powder |
132°C | |
C5HCl3N2O | |
1 g | |
KVJIRFGNHAAUNQ-UHFFFAOYSA-N | |
2,4,6-trichloropyrimidine-5-carbaldehyde | |
10932746 | |
≥98.0% (GC) | |
2,4,6-Trichloro-5-pyrimidinecarboxaldehyde |
Safety and Handling
TSCA : No