missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
GREENER_CHOICE

CAS: 191171-55-8 | C12H18BNO2 | 219.09 g/mol

$527.77 - $527.77

Chemical Identifiers

CAS 191171-55-8
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02179448
InChI Key ZCJRWQDZPIIYLM-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-ylShow MoreShow Less
PubChem CID 2734652
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367560050
SDS View Documents
Thermo Scientific Chemicals
367560050
5 g Glass bottle
Each for $527.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

Recommended Storage : product may turn dark on exposure to air

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only