missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT

CAS: 513-81-5 | C6H10 | 82.15 g/mol

Supplier:  Thermo Scientific Chemicals 163430250

 View more versions of this product

Catalog No. AC163430250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 3-Dimethyl-1, 3-butadiene
513-81-5
Colorless
68.0°C to 69.0°C
Authentic
Glass bottle
1.438
MFCD00008595
01, III, 991
0.726
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
CC(=C)C(C)=C
82.15
10566
98%
Stabilized with BHT
-76.0°C
0.7260g/mL
−20°C
97.5% min. (GC)
C6H10
H2C=C(CH3)C(CH3)=CH2
25 g
01,276; 13,45
15, 3260
SDJHPPZKZZWAKF-UHFFFAOYSA-N
2,3-dimethylbuta-1,3-diene
100ppm BHT
82.13
Liquid

Chemical Identifiers

513-81-5
82.15
SDJHPPZKZZWAKF-UHFFFAOYSA-N
10566
CC(=C)C(C)=C
C6H10
MFCD00008595
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
2,3-dimethylbuta-1,3-diene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

EINECSNumber : 208-172-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only