missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ BIO-CERT™ CELL CULTURE QUALITY 2-in-1 Culture Inserts for 12-well Plates, 8 μL Pore Size, PET Membrane

Sterile inserts for 12-well plates, featuring cellGrade™ plus surface treatment for uniform cell growth. Designed with feet for standing or a removable hook for hanging, offering versatile use in cell culture applications.
$1362.38 - $1940.47
Specifications
Barcode | No Barcode |
---|---|
Breakapart | No |
Certifications/Compliance | Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL |
Certified | Yes |
Lid | Without Lid |
Description
- Suitable for 2D and 3D cell cultures
- TC treated with cellGrade™ plus surface treatment for enhanced cell growth
- Highly transparent PET membrane ideal for imaging
- Sterile according to ISO 11137, SAL 10-6
- Produced in a Class 8 cleanroom per ISO 14644-1
- Non-cytotoxic, meeting ISO 10993-5 standards
- BIO-CERT™ CELL CULTURE QUALITY means inserts are free of pyrogens, endotoxins, human DNA, RNase, and DNase
- Diameter size of 13.3 mm for an ideal fit of 12-well plates
Specifications
No Barcode | |
Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL | |
Without Lid | |
1.38 cm2 | |
1 x 12 | |
8 μm | |
Clear | |
Microplate Insert | |
Polystyrene | |
12 | |
Sterile | |
Round, Flat Bottom (F-well) | |
2D and 3D cell culture, co-culture, migration, chemotaxis, TEER assays, and air-lift culture | |
Clear | |
13.3 mm |
No | |
Yes | |
Ambient | |
PET | |
Not Autoclavable | |
cellGrade plus | |
Single-use | |
12-well plate | |
1000 μL | |
300 to 1000 μL | |
cellGrade Plus | |
Yes | |
Yes | |
11 mm |