Learn More
(1-Ethoxycyclopropoxy)trimethylsilane, 97%

CAS: 27374-25-0 | C8H18O2Si | 174.32 g/mol
$527.47 - $527.47
Chemical Identifiers
CAS | 27374-25-0 |
---|---|
Molecular Formula | C8H18O2Si |
Molecular Weight (g/mol) | 174.32 |
MDL Number | MFCD00074986 |
InChI Key | BZMMRNKDONDVIB-UHFFFAOYSA-N |
Synonym | 1-ethoxycyclopropoxy trimethylsilane, cyclopropanone ethyl trimethylsilyl acetal, 1-ethoxycyclopropyloxy trimethylsilane, 1-ethoxy-1-trimethylsiloxylcyclopropane, 1-ethoxycyclopropyl oxy-trimethylsilane, 1-ethoxycyclopropyl-oxy trimethylsilane, silane, 1-ethoxycyclopropyl oxy trimethyl, 1-ethoxycyclopropyl oxy trimethylsilan, 1-ethoxycyclopropyl oxy trimethylsilaneShow More |
PubChem CID | 2734686 |
IUPAC Name | (1-ethoxycyclopropyl)oxy-trimethylsilane |
SMILES | CCOC1(CC1)O[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC368900250
|
Thermo Scientific Chemicals
368900250 |
25 g | Glass Bottle |
Each for $527.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27374-25-0 | |
174.32 | |
BZMMRNKDONDVIB-UHFFFAOYSA-N | |
2734686 | |
CCOC1(CC1)O[Si](C)(C)C |
C8H18O2Si | |
MFCD00074986 | |
1-ethoxycyclopropoxy trimethylsilane, cyclopropanone ethyl trimethylsilyl acetal, 1-ethoxycyclopropyloxy trimethylsilane, 1-ethoxy-1-trimethylsiloxylcyclopropane, 1-ethoxycyclopropyl oxy-trimethylsilane, 1-ethoxycyclopropyl-oxy trimethylsilane, silane, 1-ethoxycyclopropyl oxy trimethyl, 1-ethoxycyclopropyl oxy trimethylsilan, 1-ethoxycyclopropyl oxy trimethylsilaneShow More | |
(1-ethoxycyclopropyl)oxy-trimethylsilane |
Specifications
27374-25-0 | |
100.0 | |
0.867g/mL | |
99% | |
C8H18O2Si | |
CH5OC3H4OSi(CH3)3 | |
25 g | |
BZMMRNKDONDVIB-UHFFFAOYSA-N | |
(1-ethoxycyclopropyl)oxy-trimethylsilane | |
2734686 | |
≥98.5% | |
(1-Ethoxycyclopropoxy)trimethylsilane, 99% |
98.5 | |
Colorless | |
Authentic | |
Glass Bottle | |
1.4070 to 1.4090 (20°C, 589nm) | |
MFCD00074986 | |
1-ethoxycyclopropoxy trimethylsilane, cyclopropanone ethyl trimethylsilyl acetal, 1-ethoxycyclopropyloxy trimethylsilane, 1-ethoxy-1-trimethylsiloxylcyclopropane, 1-ethoxycyclopropyl oxy-trimethylsilane, 1-ethoxycyclopropyl-oxy trimethylsilane, silane, 1-ethoxycyclopropyl oxy trimethyl, 1-ethoxycyclopropyl oxy trimethylsilan, 1-ethoxycyclopropyl oxy trimethylsilane | |
CCOC1(CC1)O[Si](C)(C)C | |
174.32 | |
174.52 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only