missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-4-eth-1-ynylbenzene, 97%, Thermo Scientificâ„¢

Chemical Identifiers

CAS 79887-09-5
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00173885
InChI Key ZVWWYEHVIRMJIE-UHFFFAOYSA-N
Synonym 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene
PubChem CID 2775132
IUPAC Name 1-butyl-4-ethynylbenzene
SMILES CCCCC1=CC=C(C=C1)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09904EA
SDS View Documents
Thermo Scientific Chemicals
BTB09904EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09904EB
SDS View Documents
Thermo Scientific Chemicals
BTB09904EB
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'