Learn More
1-Allylimidazole, 97%
CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol
$93.33 - $93.33
Chemical Identifiers
CAS | 31410-01-2 |
---|---|
Molecular Formula | C6H8N2 |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00037894 |
InChI Key | XLXCHZCQTCBUOX-UHFFFAOYSA-N |
Synonym | 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #Show More |
PubChem CID | 35794 |
IUPAC Name | 1-prop-2-enylimidazole |
SMILES | C=CCN1C=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC335170250
|
Thermo Scientific Chemicals
335170250 |
25 g | Glass bottle |
Each for $93.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31410-01-2 | |
108.14 | |
XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
35794 | |
C=CCN1C=CN=C1 |
C6H8N2 | |
MFCD00037894 | |
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #Show More | |
1-prop-2-enylimidazole |
Specifications
31410-01-2 | |
100.0 | |
1.0000g/mL | |
101°C | |
96% min. (GC) | |
C6H8N2 | |
MFCD00037894 | |
1 | |
XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
1-prop-2-enylimidazole | |
35794 | |
97% | |
1-Allylimidazole, 97% |
96.0 | |
Yellow to Orange | |
212.0°C to 213.0°C | |
Authentic | |
Glass bottle | |
1.5030 to 1.5050 | |
25 g | |
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # | |
C=CCN1C=CN=C1 | |
108.14 | |
108.14 | |
Viscous Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 250-618-1
TSCA : TSCA
RUO – Research Use Only