missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diglycidyl ether, 96%

CAS: 2425-79-8 | C10H18O4 | 202.25 g/mol

$522.53 - $522.53

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1263218
View Documents
Thermo Scientific Chemicals
L1263218
50 g N/A
Only null left
 
Description

Description

1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

Solubility
Soluble in water (55,6 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

2425-79-8
202.25
SHKUUQIDMUMQQK-UHFFFAOYNA-N
17046
C(CCOCC1CO1)COCC1CO1
C10H18O4
MFCD00005146
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane

Specifications

2425-79-8
157°C to 158°C (11 mmHg)
C10H18O4
MFCD00005146
115238
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
SHKUUQIDMUMQQK-UHFFFAOYNA-N
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
17046
96%
1.049
140°C (284°F)
1.455
50 g
Hygroscopic
Soluble in water (55,6g/L at 20°C).
C(CCOCC1CO1)COCC1CO1
202.25
202.25
1,4-Butanediol diglycidyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H312-H332-H315-H319-H317
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c

H302+H312+H332-H315-H317-H318

EINECSNumber : 219-371-7

RTECSNumber : EJ5100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only