missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,2,2-Pentafluoro-4-iodobutane 98.0+%, TCI America™
Supplier: TCI America P181725G
Specifications
1,1,1,2,2-Pentafluoro-4-iodobutane | |
Yellow | |
C4H4F5I | |
25 g | |
PPXVPFPTMXFUDE-UHFFFAOYSA-N | |
1,1,1,2,2-pentafluoro-4-iodobutane | |
110092 | |
≥98.0% (GC) |
40723-80-6 | |
100°C | |
MFCD00155910 | |
3,3,4,4,4-Pentafluorobutyl Iodide | |
C(CI)C(C(F)(F)F)(F)F | |
273.972 | |
273.97 | |
Liquid |
Chemical Identifiers
40723-80-6 | |
273.972 | |
PPXVPFPTMXFUDE-UHFFFAOYSA-N | |
110092 | |
C(CI)C(C(F)(F)F)(F)F |
C4H4F5I | |
MFCD00155910 | |
3,3,4,4,4-Pentafluorobutyl Iodide | |
1,1,1,2,2-pentafluoro-4-iodobutane |
Safety and Handling
TSCA : Yes