Get Synbiosis Spiral Plate Software with Colony Counter Purchase
New Lab Start-up Program
When you purchase a Synbiosis™ ProtoCOL3 Plus Colony Counter and Zone Reader, you’ll receive a Synbiosis ProtoCOL3 Spiral Plate Software Module at no additional cost. To take advantage of this offer, order using the catalog numbers listed, log in to the Redemption Center, and complete the form to receive your software.
Features and Benefits
The Synbiosis ProtoCOL3 Plus is an all-in-one unit for counting colonies and measuring inhibition zones. Use the optional modules to extend its capabilities to reading spiral plates, OPKA, SBA, multi-sector, multi-well, Ames, and antibiotic susceptibility testing.
Get Spiral Plate Software with Colony Counter Purchase
Redeem Now
Promotion: 657
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Product
    Catalog Number Description Price Quantity & Availability  
Success
01257311 Details
GREENER_CHOICE
Synbiosis™ ProtoCOL 3 Includes: Integrated 1.4 MP CCD Camera and Mounted PC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Product Type Automated Colony Counter and Zone Measurement System
Includes Integrated 1.4 MP CCD Camera and Mounted PC
Product Line Synbiosis™
For Use With (Application) Automatic colony counting and zone measuring
Model ProtoCOL 3 Plus
Display Type LED
Dimensions (L x W x H) 500 x 400 x 700 mm
Material Injection-molded Resin
Weight (Metric) 30 kg
Sample Size 30 to 150 mm
Outlet Style Standard USA Type B Plug
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Synbiosis™ ProtoCOL 3 Software Module for Spiral Plate Measurement