Milli-Q IQ 7000: Ultrapure Water With a Touch of Intelligence
MilliporeSigma has been the partner of choice for water purification systems and services for half a century. Now, they’ve released their seventh generation of ultrapure water systems: the Milli-Q IQ 7000. The new system is the first in an offering that replaces the Milli-Q Advantage A10 systems.
Developed with You in Mind
Designed around your feedback, the Milli-Q IQ 7000 helps make your work in the lab easier and more efficient than ever. The system consistently provides water purity to the trace contaminant level, helping you avoid the uncertainty and misinterpreted results that can lead to repeating costly, time-consuming analyses. With the Milli-Q IQ 7000, you can have full confidence in your water.
An Ergonomic Design for Improved Efficiency
The system’s new Q-POD dispenser offers superior flexibility and intuitive controls. Just twist the dispensing wheel to deliver water at the flow rate you need — from drop-by-drop to two liters per minute. Or, select dispensing options from the Q-POD Home screen and check water quality parameters onscreen even during dispensing.
Each system can be used with up to four Q-POD dispensers installed in the same lab or in two or more adjacent spaces. The compact water production unit can easily be mounted to a wall or under a bench, and the customizable-length system cables are enclosed in protective sheaths. Choose from a range of POD-Pak cartridge filters to produce the precise water quality you need.
Intuitive Graphic Interface
Everyday interactions with the Milli-Q IQ 7000 are performed on the high-definition digital touchscreen. Navigation is intuitive, and information is organized to provide an efficient user experience. The system can also be connected to lab networks through an integrated data management system for quick and easy access to information.
The system puts help for daily use right at your fingertips, and will alert you with an onscreen message when consumables need to be replaced. It even automatically provides step-by-step guidance for changing the cartridges.
Novel Mercury-Free UV Lamps
The Milli-Q IQ 7000 pioneers the use of environmentally friendly, mercury-free UV lamps. It also features a 23% smaller system footprint as well as IPAK Meta and IPAK Quanta cartridges that are 33% smaller than before. With these changes, the system requires less plastic for manufacturing and fewer packaging materials to reduce shipping costs and waste volume. Online documentation reduces paper use, and the unique LabClose mode places the system in hibernation to reduce water and electricity consumption.
Precise A10 TOC Monitoring
The Milli-Q IQ 7000 delivers ultrapure water with low TOC (Total Organic Carbon) levels (≤ 2 to ≤ 5 ppb). The A10 TOC monitor is also equipped with an ech2o mercury-free UV oxidation lamp that has been redesigned for greater accuracy. (Its 0.5 – 999.9 range and precision comply with ESP and EP suitability tests.)