CAS RN 9005-64-5
Polysorbate 20, Fisher BioReagents™
Non-ionic detergent | CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
TWEEN 20, MP Biomedicals™
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polyethyleneglycol sorbitan monolaurate,Polyoxyethylenesorbitan monolaurate,Polysorbate 20,Polyoxyethylene (20) Sorbitan Monolaurate IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MilliporeSigma™ Tween™ 20 (Polyoxyethylene (20) Monolaurate)OmniPur™, Calbiochem™,
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: tween 20,polysorbate 20,polyoxyethylene sorbitan monolaurate,polyoxyethylene 20 sorbitan monolaurate,polysorbate inn,2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate,alkest tw 20,polysorbate 20 nf,polysorbate 40 nf PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MilliporeSigma™ TWEEN™ 20, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polyoxyethylene (20) sorbitan Monolaurate PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
TWEEN™ 20 Detergent, Molecular biology grade, MilliporeSigma™
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polysorbate 20 PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polyoxyethylene 20 Sorbitan Monolaurate, BAKER™, J.T. Baker™
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20, Vegetable, Multi-Compendial, N.F., J.T. Baker™
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N PubChem CID: 443314 SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: tween 20,polysorbate 20,polyoxyethylene sorbitan monolaurate,polyoxyethylene 20 sorbitan monolaurate,polysorbate inn,2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate,alkest tw 20,polysorbate 20 nf,polysorbate 40 nf PubChem CID: 443314 IUPAC Name: 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20
CAS: 9005-64-5 Molecular Formula: C26H50O10 Synonym: Polyoxyethylene(20)sorbitan monolaurate
Tween 20™, Ultrapure, Thermo Scientific Chemicals
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polyoxyethylene sorbitan monolaurate PubChem CID: 443314 SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20, Electrophoresis Reagent
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polyoxyethylene sorbitan monolaurate; Tween™ 20 PubChem CID: 443314 IUPAC Name: 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N PubChem CID: 443314 SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20 MP Biomedicals
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polysorbate 20,Glycosperse L-20X,Polyoxyethylene (20) Sorbitan Monolaurate PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 20, 100g, MP Biomedicals
CAS: 9005-64-5 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N Synonym: Polyoxyethylenesorbitan monolaurate PubChem CID: 443314 IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
- 1
- 2