missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tween™ 20 (Polyoxyethylene (20) Monolaurate)OmniPur™, Calbiochem™,

Catalog No. 94854L
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94854L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 94854L Supplier MilliporeSigma™ Supplier No. 94854L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nonionic detergent suited for solubilization of membrane proteins, enzymology, analytical, and diagnostic work, as well as chromatography and electrophoresis

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
PubChem CID 443314
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Colorless to Yellow
Quantity 4 L
Physical Form Liquid
Solubility Information Water: Soluble
Formula Weight 1227.72
Chemical Name or Material Tween 20
Grade Molecular biology grade
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.