missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, 100g, MP Biomedicals

Catalog No. ICN806576
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN806576 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN806576 Supplier MP Biomedicals Inc Supplier No. 04806576

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polysorbate 20 is used as emulsifying agent for the preparation of stable oil-in-water emulsions in pharmaceuticals, cosmetics, cleaning compounds. Polysorbate 20 has been used in pre-extraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins).

Polysorbate 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%. It can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. It is a nonionic detergent widely used in biochemical applications.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylenesorbitan monolaurate
PubChem CID 443314
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 100 g
Chemical Name or Material Polysorbate 20