missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™

Catalog No. 02003672
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Steel Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003672 4 L Narrow Mouth Amber Glass Bottle
02003671 20 mL Narrow Mouth Amber Glass Bottle
02003669 200 mL Narrow Mouth Amber Glass Bottle
02003665 1 L Narrow Mouth Amber Glass Bottle
02003664 19 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02003672 Supplier Avantor Supplier No. 411603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

pH 5.0 to 7.0
Quantity 4 L
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.11kg
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 20