missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ TWEEN™ 20 Detergent, Calbiochem™,

Catalog No. 655205250ML
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
655205250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 655205250ML Supplier MilliporeSigma™ Supplier No. 655205250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-ionic detergent used for immunoprecipitation and the solubilization of membrane-bound proteins.

Chemical Identifiers

CAS 9005-64-5

Specifications

Color Yellow
Quantity 250 mL
Format Glass Bottle