missing translation for 'onlineSavingsMsg'
Learn More

TWEEN 20, MP Biomedicals™

Catalog No. MP1TWEEN201
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1TWEEN201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1TWEEN201 Supplier MP Biomedicals Inc Supplier No. 11TWEEN201
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TWEEN 20, Molecular biology reagent, Viscous pale yellow liquid

  • TWEEN 20 is a nonionic detergent widely used in biochemical applications, such as emulsifying agents for the preparation of stable oil-inwater emulsions.
  • It has been used in preextraction of membranes to remove peripheral proteins.
  • TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%.
  • TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. TWEEN 20 is a polyoxyethylene sorbitol ester.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyethyleneglycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Pale Yellow
Physical Form Liquid
Boiling Point >100°C (literature)
Density 1.09 - 1.12(25°C)
Quantity 500 mL
Flash Point >230°F/ >110°C (Lit.)
Refractive Index < 1.33 hPa (< 1.00 mmHg, Lit.)
Specific Gravity 1.08 - 1.13
Viscosity Brookfield Viscosity: 370-430 cps (25°C, neat, Lit.)
Water ≤3%
Solubility Information Soluble in Water(100mg/mL).
Grade Molecular Biology
Show More Show Less
RTECSNumber TR7400000
storageNote1 Store at Room temperature(15-30°C).