accessibility menu, dialog, popup

UserName

O∗O∗OORORORORORORn

CAS RN 9004-32-4

IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Synonyms: cellulose gum
Molecular Weight (g/mol): —
Molecular Formula: (C12 H14 O9 R6)n
InChi Key: DPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES: [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
Quantity 
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
Grade 
  • (5)
  • (12)
Percent Purity 
  • (11)
  • (5)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (25)
  • (12)
  • (3)
  • (3)
  • (12)
  • (3)
  • (13)
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (5)
  • (12)
  • (11)
  • (5)
  • (1)
  • (3)
  • (3)
  • (3)